4-(Dimethylamino)benzaldehyde 98%

Supplier: ALFA AESAR
Warning

A11712.0E A11712.22 A11712.36
A11712.0EEA 478 EUR
A11712.0E A11712.22 A11712.36
4-(Dimethylamino)benzaldehyde 98%
4-(Dimethylamino)benzaldehyde

Purity: 98% CAS No.: 100-10-7 Molecular Formula: C9H11NO Formula Weight: 149.19 Flash Point: 164°(327°F) Odour: Strong Boiling Point: 176-177°/17mm Beilstein Reference: 606802 Merck Reference: 14,3230 MDL No.: MFCD00003381

Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176…177 °C (17 mmHg)
Melting Pt: 73…75 °C
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: 164 °C (327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 14,3230
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=a978f8cb2be9fb6dfa0aa34ce0038883c864820b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR