Stearic acid ≥98%

Supplier: ALFA AESAR
Warning

A12244.06 A12244.14 A12244.22
A12244.06EA 37.6 EUR
A12244.06 A12244.14 A12244.22
Stearic acid ≥98%
Stearic acid

Purity: 98% CAS No.: 57-11-4 Molecular Formula: CH3(CH2)16CO2H Formula Weight: 284.48 Flash Point: 196°(384°F) Physical Form: Flakes or powder Density: 0.941 Appearance: White Odour: Odourless Melting Point: 69-72° Boiling Point: 232°/15mm Beilstein Reference: 608585 Merck Reference: 14,8804 MDL No.: MFCD00002752


A saturated fatty acid

Formula: C₁₈H₃₆O₂
Boiling Pt: 232 °C (15 mmHg)
Melting Pt: 69…72 °C
Density: 0,941
Flash Pt: 196 °C (384 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 14,8804
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR