5-(Hydroxymethyl)-2-furaldehyde ≥97%

Supplier: ALFA AESAR
Warning

A12475.03 A12475.06 A12475.14
A12475.03EA 71.8 EUR
A12475.03 A12475.06 A12475.14
5-(Hydroxymethyl)-2-furaldehyde ≥97%
5-(Hydroxymethyl)-2-furaldehyde

Purity: 97% CAS No.: 67-47-0 Molecular Formula: C6H6O3 Formula Weight: 126.11 Storage Temperature: 2°C to 8°C Flash Point: 79°(174°F) Refractive Index: 1.5620 Melting Point: 32-35° Boiling Point: 114-116°/1mm Beilstein Reference: 110889 Merck Reference: 14,4832 MDL No.: MFCD00003234

Formula: C₆H₆O₃
Boiling Pt: 114…116 °C (1 mmHg)
Melting Pt: 32…35 °C
Density: 1,206
Flash Pt: 79 °C (174 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003234
CAS Number: 67-47-0
EINECS: 200-654-9
Merck Index: 14,4832

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR