1-Octanesulphonic acid sodium salt ≥99%

Supplier: ALFA AESAR
Warning

A14292.14 A14292.06 A14292.22
A14292.14EA 166 EUR
A14292.14 A14292.06 A14292.22
1-Octanesulphonic acid sodium salt ≥99%
1-Octanesulphonic acid sodium salt

Purity: 99% CAS No.: 5324-84-5 Molecular Formula: CH3(CH2)7SO3Na Formula Weight: 216.28 Melting Point: 300° Beilstein Reference: 3574241


Reagent for HPLC analysis of peptides and proteins

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=92242d1e72ac2fd7b5d0878e64a4a264275fedea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR