3-Chloropropene ≥98% stabilised

Supplier: ALFA AESAR
Danger


Stabilization: Stabilised with propylene oxide

A14330.0F A14330.AP
A14330.0FEA 122 EUR
A14330.0F A14330.AP
3-Chloropropene ≥98% stabilised
3-Chloropropene

Stabilized with propylene oxide


Purity: 98% CAS No.: 107-05-1 Molecular Formula: C3H5Cl Formula Weight: 76.53 Storage Temperature: 2°C to 8°C Flash Point: -28°(-18°F) Refractive Index: 1.4155 Odour: Sweet Melting Point: -135° Boiling Point: 44-46° Beilstein Reference: 635704 Merck Reference: 14,289 UN No.: 1100 MDL No.: MFCD00000984

Formula: C₃H₅Cl
Boiling Pt: 44…46 °C
Melting Pt: –135 °C
Density: 0,939
Flash Pt: –28 °C (–18 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000984
CAS Number: 107-05-1
EINECS: 203-457-6
UN: 1100
ADR: 3,I
Merck Index: 14,289

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR