Actinomycin D ≥92% (by HPLC)

A1489.0005
A1489.0005EA 0 EUR
A1489.0005
Actinomycin D ≥92% (by HPLC)
Actinomycin D
Formula: C₆₂H₈₆N₁₂O₁₆
MW: 1255.5 g/mol
MDL Number: MFCD00005033
CAS Number: 50-76-0
UN: 3462
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR