1-(4-Iodophenyl)pyrrole ≥97%

Supplier: ALFA AESAR

A15305.03 A15305.06
A15305.03EA 47.5 EUR
A15305.03 A15305.06
1-(4-Iodophenyl)pyrrole ≥97%
1-(4-Iodophenyl)pyrrole

Purity: 97% CAS No.: 92636-36-7 Molecular Formula: C10H8IN Formula Weight: 269.09 Melting Point: 128-133° Beilstein Reference: 7915717

Formula: C₁₀H₈IN
Melting Pt: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00052399
CAS Number: 92636-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR