Sodium sulphate decahydrate ≥99%

Supplier: ALFA AESAR

A15702.0B A15702.0G A15702.0I
A15702.0BEA 60.5 EUR
A15702.0B A15702.0G A15702.0I
Sodium sulphate decahydrate ≥99%
Sodium sulphate decahydrate

Purity: 99% CAS No.: 7727-73-3 Molecular Formula: Na2SO4.10H2O Formula Weight: 322.19 (142.04anhy) Odour: Odourless pH: 5.2-8.0 Melting Point: ca 32° Merck Reference: 14,8680 MDL No.: MFCD00149189

Formula: Na₂SO₄·10H₂O
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: ∼32 °C
Density: 1,460
Storage Temperature: Ambient
MDL Number: MFCD00149189
CAS Number: 7727-73-3
EINECS: 231-820-9
Merck Index: 14,8680

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR