Potassium hexacyanoferrate(II) trihydrate ≥98%

Supplier: ALFA AESAR

A15736.0E A15736.22 A15736.36
A15736.0EEA 255 EUR
A15736.0E A15736.22 A15736.36
Potassium hexacyanoferrate(II) trihydrate ≥98%
Potassium hexacyanoferrate (II) trihydrate

Purity: 98+% CAS No.: 14459-95-1 Molecular Formula: K4Fe(CN)6.3H2O Formula Weight: 422.39 (368.36anhy) Odour: Odourless pH: 9.5 Melting Point: 69-71° Merck Reference: 14,7631 MDL No.: MFCD00167023

Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 69…71 °C
Density: 1.85
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR