Glycerine ≥99%

Supplier: ALFA AESAR
Warning

A16205.0D A16205.0F A16205.AP
A16205.0DEA 338 EUR
A16205.0D A16205.0F A16205.AP
Glycerine ≥99%
Glycerine

Purity: 99+% CAS No.: 56-81-5 Molecular Formula: C3H8O3 Formula Weight: 92.09 Flash Point: 160°(320°F) Physical Form: Viscous liquid Density: 1.260 Refractive Index: 1.4740 Sensitivity: Hygroscopic Appearance: Colourless Odour: Odourless Melting Point: 18° Boiling Point: 182°/20mm Beilstein Reference: 635685 Merck Reference: 14,4484 MDL No.: MFCD00004722

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 182 °C (20 mmHg)
Melting Pt: 18 °C
Density: 1.260
Flash Pt: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=481dd90ecbdec73cd4020432d3356161ebfe7681&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR