2-Nitrophenyl octyl ether ≥98%

Supplier: ALFA AESAR

A17320.14 A17320.06
A17320.14EA 236 EUR
A17320.14 A17320.06
2-Nitrophenyl octyl ether ≥98%
2-Nitrophenyl octyl ether

Purity: 98% CAS No.: 37682-29-4 Molecular Formula: C14H21NO3 Formula Weight: 251.32 Flash Point: 110°(230°F) Refractive Index: 1.5110 Boiling Point: 163-165°/2mm Beilstein Reference: 2529905 MDL No.: MFCD00014693

Formula: C₁₄H₂₁NO₃
MW: 251.32 g/mol
Boiling Pt: 163…165 °C (2 mmHg)
Density: 1.04
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014693
CAS Number: 37682-29-4
EINECS: 253-623-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR