D-(-)-Ribose ≥98%

Supplier: ALFA AESAR

A17894.09 A17894.18 A17894.30
A17894.09EA 32.5 EUR
A17894.09 A17894.18 A17894.30
D-(-)-Ribose ≥98%
D(-)-Ribose

Purity: 98% CAS No.: 50-69-1 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 82-86° Beilstein Reference: 1723081 Merck Reference: 14,8204 Optical Rotation: -20° (c=10 in water, 28h) MDL No.: MFCD00135453

Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 82…86 °C
Density: 0,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 14,8204

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR