Anti-His6-FAP Rabbit Polyclonal Antibody

Supplier: Abgent
AP12541A
ABGEAP12541AEA 354 EUR
ABGEAP12541A
Anti-His6-FAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: His6-FAP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR