Avantor® ACE® C18-AR, HPLC Columns, Microbore, 2 µm
Supplier: Advanced Chromatography Technologies
ACE-109-05005SEA 1460 EUR
ACE-109-05005S
Avantor® ACE® C18-AR, HPLC Columns, Microbore, 2 µm
Chromatography Columns
The Avantor® ACE® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and
More Product Information