Acetophenone 98%, pure

Supplier: ACROS ORGANICS
Warning

102410010 102410025 102410050 102412500 102410100
ACRO102410010EA 56.7 EUR
ACRO102410010 ACRO102410025 ACRO102410050 ACRO102412500 ACRO102410100
Acetophenone 98%, pure
Acetophenone
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: 19.6 °C
Density: 1.028 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
Merck Index: 13,00074

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20°C, 589 nm)
GC ≥97.5 %
Visual transition interval from pH 9.4 (orange-brown) to pH 12.0 (violet)
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR