3-Mercapto-1,2-propanediol 90% 90% in aqueous solution

Supplier: ACROS ORGANICS
Danger

125520250 125521000 125525000
ACRO125520250EA 51.3 EUR
ACRO125520250 ACRO125521000 ACRO125525000
3-Mercapto-1,2-propanediol 90% 90% in aqueous solution
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
Boiling Pt: 118 °C (5 mmHg)
Density: 1,23 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
UN: 2810

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous solution
Infrared spectrum Conforms
Titration Iodimetric ≥88 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR