Methyl-4-hydroxybenzoate 99%

Supplier: ACROS ORGANICS
Warning

126961000 126965000 126960025 126960100
ACRO126961000EA 42.2 EUR
ACRO126961000 ACRO126965000 ACRO126960025 ACRO126960100
Methyl-4-hydroxybenzoate 99%
Methyl 4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 298.6 °C (760 mmHg)
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=84321052052f4be2a53d8d7ebac7fabe6fc5a435&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 128 °C
HPLC ≥98.5 %
Residue after ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR