2-Thiohydantoin 98%

Supplier: ACROS ORGANICS
Warning

138760050 138760250 138761000
ACRO138760050EA 36.4 EUR
ACRO138760050 ACRO138760250 ACRO138761000
2-Thiohydantoin 98%
2-Thiohydantoin
Formula: C₃H₄N₂OS
MW: 116.14 g/mol
Melting Pt: 229…231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005277
CAS Number: 503-87-7
EINECS: 207-977-4

Order Now

Specification Test Results

Appearance Cream to yellow to beige to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR