o-Toluidine 99%

Supplier: ACROS ORGANICS
Danger

139080025 139085000 139080100
ACRO139080025EA 164 EUR
ACRO139080025 ACRO139085000 ACRO139080100
o-Toluidine 99%
o-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 199…200 °C (760 mmHg)
Melting Pt: –23 °C
Density: 1 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007730
CAS Number: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1 II
Merck Index: 12,09674

Order Now

Specification Test Results

Appearance Clear yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5710 to 1.5730 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR