Diphenylamine 99%, pure

Supplier: ACROS ORGANICS
Danger

150740010 150740050 150740051 150741000
ACRO150740010EA 178 EUR
ACRO150740010 ACRO150740050 ACRO150740051 ACRO150741000
Diphenylamine 99%, pure
Diphenylamine
Formula: (C₆H₅)₂NH
Boiling Pt: 302 °C (760 mmHg)
Melting Pt: 52…54 °C
Density: 1,165 g/cm³ (20 °C)
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
Merck Index: 13,03349
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=7573b36ed017a251a44814143c7c7e4077d3a7b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to beige Flakes
Infrared spectrum Conforms
Melting point 52 to 55 °C
GC ≥98.5 %
Free acid ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR