N,N'-Diethylethylenediamine 95%

Supplier: ACROS ORGANICS
Danger

155520050 155520250
ACRO155520050EA 42.4 EUR
ACRO155520050 ACRO155520250
N,N'-Diethylethylenediamine 95%
N,N'-Diethylethylenediamine
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 152…154 °C (760 mmHg)
Melting Pt: –11 °C
Density: 0.811 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009033
CAS Number: 111-74-0
UN: 2685
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4315 to 1.4335 (20°C, 589 nm)
GC ≥94 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR