2,6-Di-tert-butyl-p-benzoquinone 98%

Supplier: ACROS ORGANICS
Warning

167340010 167340050 167340250
ACRO167340010EA 24.7 EUR
ACRO167340010 ACRO167340050 ACRO167340250
2,6-Di-tert-butyl-p-benzoquinone 98%
2,6-Di-tert-butyl-p-benzoquinone
Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Boiling Pt: 75…80 °C (.7 mmHg)
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00001601
CAS Number: 719-22-2

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 64 to 70 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR