2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid 99+% for biochemistry

Supplier: ACROS ORGANICS
Warning

172580250 172581000 172585000
ACRO172580250EA 41 EUR
ACRO172580250 ACRO172581000 ACRO172585000
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid 99+% for biochemistry
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Formula: C₆H₁₅NO₅S
Boiling Pt: 405 °C (1013 hPa)
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00007533
CAS Number: 10191-18-1
EINECS: 233-465-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (100°C, 2 hrs)
Sulfated ash ≤0.1 %
Solubility (1 M in water) Clear colorless
UV at 280 nm A: ≤0.080
Titration with CHA ≥99.0 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR