Sodium gluconate 98%

Supplier: ACROS ORGANICS

181390010 181390025 181390050 181390100
ACRO181390010EA 74.8 EUR
ACRO181390010 ACRO181390025 ACRO181390050 ACRO181390100
Sodium gluconate 98%
Sodium gluconate
Formula: HOH₂C(CH(OH))₄COONa
MW: 218.14 g/mol
Melting Pt: 206…209 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
pH 6.5 to 7.5 (10 % solution)
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤100 ppm
Lead (Pb) ≤10 ppm
Reducing substances ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR