Ammonium sulphate 99.9995% (trace metals basis)

Supplier: ACROS ORGANICS

194150500 194152500
ACRO194150500EA 108 EUR
ACRO194150500 ACRO194152500
Ammonium sulphate 99.9995% (trace metals basis)
Ammonium sulphate
Formula: (NH₄)₂SO₄
Melting Pt: 280 °C
Density: 1,78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤5 ppm
Gravimetry 23.7 to 24.9 % (S)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR