Rhodium 5% on activated carbon, dry, powder, unreduced

Supplier: ACROS ORGANICS

195440050 195440250 195440010
ACRO195440050EA 827 EUR
ACRO195440050 ACRO195440250 ACRO195440010
Rhodium 5% on activated carbon, dry, powder, unreduced
Rhodium
Formula: Rh
Boiling Pt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960 °C
Density: 12,41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6

Order Now

Specification Test Results

Appearance Grey-black to black Powder
Assay 4.5 to 5.5 % (Rh)
Particle Size/Sieve analysis <128 um: 99%
Additional info 128 um: 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR