Zinc 98+% stable (acc. to UN classification class 4), dust

Supplier: ACROS ORGANICS
Warning

198340010 198340025 198341000 198340100 198340250
ACRO198340010EA 63.5 EUR
ACRO198340010 ACRO198340025 ACRO198341000 ACRO198340100 ACRO198340250
Zinc 98+% stable (acc. to UN classification class 4), dust
Zinc

Stable acc. to UN classification class 4

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 906 °C (760 mmHg)
Melting Pt: 419 °C
Density: 7.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9 III
Merck Index: 12,10255

Order Now

Specification Test Results

Appearance Grey Powder
Particle Size/Sieve analysis ≤0.1 % Residue on 250 µm(+60 mesh) sieve
Zinc (Zn) ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR