Lithium nitrate 99.999% (trace metals basis)

Supplier: ACROS ORGANICS
Warning

212540050 212540250 212541000
ACRO212540050EA 71.9 EUR
ACRO212540050 ACRO212540250 ACRO212541000
Lithium nitrate 99.999% (trace metals basis)
Lithium nitrate
Formula: LiNO₃
MW: 68.95 g/mol
Boiling Pt: 600 °C (760 mmHg)
Melting Pt: 251 °C
Density: 2.38 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
UN: 2722
ADR: 5.1 III
Merck Index: 13,05558

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Total trace metal impurities ≤10 ppm
Lithium (Li) 9 to 11 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR