Copper(I) bromide dimethyl sulphide complex 99%

Supplier: ACROS ORGANICS

213180100 213180250 213181000
ACRO213180100EA 114 EUR
ACRO213180100 ACRO213180250 ACRO213181000
Copper(I) bromide dimethyl sulphide complex 99%
Copper (I) bromide dimethyl sulphide complex
Formula: C₂H₆BrCuS
Melting Pt: 132 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043295
CAS Number: 54678-23-8
EINECS: 259-292-5

Order Now

Specification Test Results

Appearance Off-white to beige to green to grey Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Iodometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR