Cadmium(II) chloride, anhydrous 99% for analysis

Supplier: ACROS ORGANICS
Danger

219141000 219145000
ACRO219141000EA 144 EUR
ACRO219141000 ACRO219145000
Cadmium(II) chloride, anhydrous 99% for analysis
Cadmium (II) chloride
Formula: CdCl₂
Boiling Pt: 967 °C (1013 hPa)
Melting Pt: 568 °C
Density: 4,05 g/cm³ (20 °C)
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 12,01653

Order Now

Specification Test Results

Appearance White Powder
Titration Argentometric ≥98.5 % (On dry substance)
Nitrate and Nitrite (as NO3) ≤0.003 %
Sulfate (SO4) ≤100 ppm
Ammonium (NH4) ≤100 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤50 ppm
Insoluble matter ≤0.01 %
Subst not pptd by H2S ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR