Ruthenium(III) chloride oxide ≥85%, pure

Supplier: ACROS ORGANICS
Warning

279710010 279710050
ACRO279710010EA 145 EUR
ACRO279710010 ACRO279710050
Ruthenium(III) chloride oxide ≥85%, pure
Ruthenium(III) chloride oxide
Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
Melting Pt: >500 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now

Specification Test Results

Appearance Dark red or purple Powder
Elemental Analysis 34 to 39 % (Ru)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR