Acetic acid ≥99,6% 99.6% for analysis

Supplier: ACROS ORGANICS
Danger

295320010 295320025 295320100
ACRO295320010EA 58 EUR
ACRO295320010 ACRO295320025 ACRO295320100
Acetic acid ≥99,6% 99.6% for analysis
Acetic acid
Formula: H₃CCOOH
Melting Pt: 16…16,5 °C
Density: 1,048 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=031896546f0eb78526a541f0af4636e3991e9f8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.2 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Titration with NaOH ≥99.5 %
Chloride (Cl) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Arsenic (As) ≤0.5 ppm
Cadmium (Cd) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
KMnO4-reducing substance Passes test
K2Cr2O7-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR