(4-Nitrophenyl)-β-D-glucopyranoside 99%

Supplier: ACROS ORGANICS

303110010 303110050 303110250
ACRO303110010EA 112 EUR
ACRO303110010 ACRO303110050 ACRO303110250
(4-Nitrophenyl)-β-D-glucopyranoside 99%
(4-Nitrophenyl) β-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Freezer
MDL Number: MFCD00006593
CAS Number: 2492-87-7
EINECS: 219-661-3

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥98.5 % (ELSD)
Specific optical rotation -96° to -117° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR