Methyl-4-(bromomethyl)benzoate 98%

Supplier: ACROS ORGANICS
Danger

306100050 306100250 306101000
ACRO306100050EA 77.7 EUR
ACRO306100050 ACRO306100250 ACRO306101000
Methyl-4-(bromomethyl)benzoate 98%
Methyl 4-(bromomethyl)benzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130…135 °C (2 mmHg)
Melting Pt: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00032453
CAS Number: 2417-72-3
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR