2-Chloro-p-benzoquinone 95%

Supplier: ACROS ORGANICS
Warning

331220050 331220250
ACRO331220050EA 91.7 EUR
ACRO331220050 ACRO331220250
2-Chloro-p-benzoquinone 95%
2-Chloro-p-benzoquinone
Formula: C₆H₃ClO₂
MW: 142.54 g/mol
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00075260
CAS Number: 695-99-8

Order Now

Specification Test Results

Appearance Yellow to brown Crystals
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR