(4-Nitrophenyl)-α-D-glucopyranoside 99%

Supplier: ACROS ORGANICS

337150050 337150010 337150250
ACRO337150050EA 313 EUR
ACRO337150050 ACRO337150010 ACRO337150250
(4-Nitrophenyl)-α-D-glucopyranoside 99%
(4-Nitrophenyl) α-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 204…218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064088
CAS Number: 3767-28-0
EINECS: 223-189-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Solubility (2% in methanol) Clear to slightly hazy colorless to green-yellow solution
HPLC ≥98.5 %
Specific optical rotation +210° to +220° (20°C, 589 nm) (c=1, water)
Impurity ≤0.1 % (p-Nitrophenol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR