cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 97%

Supplier: ACROS ORGANICS
Warning

367500010 367500050 367502500 367505000
ACRO367500010EA 329 EUR
ACRO367500010 ACRO367500050 ACRO367502500 ACRO367505000
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 97%
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate

CAS No.: 98014-14-3

Formula: C₂₀H₁₈Cl₂N₄ORu
MW: 502.361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150005
CAS Number: 98014-14-3

Order Now

Specification Test Results

Appearance Dark green to black Powder and/or chunks
Infrared spectrum Conforms
Assay ≥96.0 % (Based on Carbon)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR