Potassium hydroxide 8 mol/l (8 N) in water, pure

Supplier: ACROS ORGANICS
Danger

380625000 380620020 380620100
ACRO380625000EA 39.7 EUR
ACRO380625000 ACRO380620020 ACRO380620100
Potassium hydroxide 8 mol/l (8 N) in water, pure
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.326 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 7.984 to 8.016 N (20°C)
Composition 448 g/l (g KOH/l H20)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR