2-Propanol 99.5%, gradient grade for HPLC

Supplier: ACROS ORGANICS
Danger

383920025 383920010
ACRO383920025EA 178 EUR
ACRO383920025 ACRO383920010
2-Propanol 99.5%, gradient grade for HPLC
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 81…83 °C (760 mmHg)
Melting Pt: –89.5 °C
Density: 0.785 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 %
GC ≥99.4 %
UV at 250 nm A: ≤0.01
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0001 meq/g
Gradient analysis :
Absorbance largest eluted peak ≤0.002 A.U.
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR