N-Methyl-2-pyrrolidone (NMP) 99+% ACS

Supplier: ACROS ORGANICS
Danger

390682500 390680010 390680025 390680100
ACRO390682500EA 64.4 EUR
ACRO390682500 ACRO390680010 ACRO390680025 ACRO390680100
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: –24 °C
Density: 1.03 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=e8ca0ad6010f87ad7cf1c890c0c8248772262ede&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.50 %
Color scale ≤50 APHA
Chloride (Cl) ≤1 ppm
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Free amine ≤0.01 % (as CH3NH2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR