Silica gel 2-5 mm, non toxic for drying, with moisture indicator (orange gel)

Supplier: ACROS ORGANICS

392030010 392030050 392032500
ACRO392030010EA 109 EUR
ACRO392030010 ACRO392030050 ACRO392032500
Silica gel 2-5 mm, non toxic for drying, with moisture indicator (orange gel)
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00077645
CAS Number: 7631-86-9

Order Now

Specification Test Results

Appearance Orange Beads
Bead size 2 to 5 mm
Color Orange (dry) to Colorless (wet)
Residual water ≤2 % (150°C)
Water adsorption ≥20 % (50% rH, 1 bar, 25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR