2-Amino-1,3-propanediol 98%

Supplier: ACROS ORGANICS
Danger

401030050 401030250
ACRO401030050EA 78.4 EUR
ACRO401030050 ACRO401030250
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol
Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: 52…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0
UN: 3259
ADR: 8 III

Order Now

Specification Test Results

Appearance White to off-white Adhering crystalline powder
Infrared spectrum Conforms
Melting point 52 to 56 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR