Guanosine 99%

Supplier: ACROS ORGANICS
Danger

411130250 411131000 411135000
ACRO411130250EA 82.8 EUR
ACRO411130250 ACRO411131000 ACRO411135000
Guanosine 99%
Guanosine
Formula: C₁₀H₁₃N₅O₅
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
EINECS: 204-227-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Titration with HClO4 ≥98.5 % (On dry substance)
Specific optical rotation -9.5° to -12.5° (20°C, 589 nm) (c=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR