Triethanolamine (trolamine) 99+%

Supplier: ACROS ORGANICS

421630010 421630025 421631000 421630250
ACRO421630010EA 112 EUR
ACRO421630010 ACRO421630025 ACRO421631000 ACRO421630250
Triethanolamine (trolamine) 99+%
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 21 °C
Density: 1,1245 g/cm³
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=c181d3e80375aec7c34e17766247484e1c3d3257&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4860 (20°C, 589 nm)
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR