Thiopicolinamide 97%

Supplier: ACROS ORGANICS
Danger

432470050 432470250
ACRO432470050EA 38.9 EUR
ACRO432470050 ACRO432470250
Thiopicolinamide 97%
Thiopicolinamide
Formula: C₆H₆N₂S
MW: 138.19 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00087576
CAS Number: 5346-38-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 134 to 140 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR