1-Propanol ACS for spectroscopy

Supplier: ACROS ORGANICS
Danger

434360010 434360025
ACRO434360010EA 74.8 EUR
ACRO434360010 ACRO434360025
1-Propanol ACS for spectroscopy
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –127 °C
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=b4a45c3a7a92d9e6aed8d1d3bcf6b139b7a0e31f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.2 %
Solubility Clear colorless
GC ≥99.5 %
UV at 220 nm A: ≤0.7
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.03 % (as C2H5CHO)
Isopropyl alcohol ≤0.05 %
Titrable acid 0.0004 meq/g
Ethanol ≤0.01 %
Methanol ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR