Methyl ethyl ketone, Technical Grade

Supplier: ACROS ORGANICS
Danger

444170050
ACRO444170050EA 66.2 EUR
ACRO444170050
Methyl ethyl ketone, Technical Grade
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –87 °C
Density: 0.806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3 II
Merck Index: 13,06097
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=7560bf484cae1b3d9ae7f8cc59b75018d61b5022&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR