Itraconazole 99%

Supplier: ACROS ORGANICS
Warning

452870050 452870250 452870000
ACRO452870050EA 147 EUR
ACRO452870050 ACRO452870250 ACRO452870000
Itraconazole 99%
Itraconazole
Formula: C₃₅H₃₈Cl₂N₈O₄
Melting Pt: 168…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00870168
CAS Number: 84625-61-6

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
Loss on drying ≤0.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR