Aliquat® 336 TG

Supplier: ACROS ORGANICS
Danger

463570025 463570010 463572500
ACRO463570025EA 254 EUR
ACRO463570025 ACRO463570010 ACRO463572500
Aliquat® 336 TG
Aliquat® 336
Formula: CH₃(CH₂)₇]₃NCH₃Cl
MW: 404.17 g/mol
Density: 0.89 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00011862
CAS Number: 63393-96-4
UN: 2922
ADR: 8 III

Order Now

Specification Test Results

Appearance Yellow to amber Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4655 to 1.4715 (20°C, 589 nm)
Assay 112.0 to 117.2 (Quaternary value)
Water ≤5 %
Titration with HClO4 88.2 to 93.0 % (≤2.0 % (Tert.amine))


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR