β-Methasone sodium phosphate

Supplier: ACROS ORGANICS
Warning

465740010 465740050
ACRO465740010EA 122 EUR
ACRO465740010 ACRO465740050
β-Methasone sodium phosphate
β-Methasone sodium phosphate
Formula: C₂₂H₂₈FO₈P·₂Na
MW: 516.41 g/mol
Melting Pt: 206°C…211°C
Storage Temperature: Refrigerator
MDL Number: MFCD00200361
CAS Number: 151-73-5
EINECS: 205-797-0

Order Now

Specification Test Results

Appearance White to almost white powder
Infrared spectrum Conforms
Water ≤ 10%
HPLC ≥ 96.0%
Specific optical rotation +98° to +105° (20°C, 589nm) (1% in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR